missing translation for 'onlineSavingsMsg'
Learn More

RNASEH2B Antibody [Alexa Fluor™ 405], Novus Biologicals™

Código de producto. 30599718 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30599718 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Product Code. 30599718 Supplier Novus Biologicals Supplier No. NBP346718AF405

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNASEH2B Polyclonal antibody specifically detects RNASEH2B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno RNASEH2B
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Alexa Fluor 405
Formulación 50mM Sodium Borate
Alias de gen ribonuclease H2, subunit B
Especie del huésped Rabbit
Inmunógeno ribonuclease H2, subunit B. (Uniprot# Q5TBB1)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 79621
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.