missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RGS7 Polyclonal antibody specifically detects RGS7 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antígeno | RGS7 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Janelia Fluor 646 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | regulator of G-protein signaling 7, regulator of G-protein signaling RGS7, regulator of G-protein signalling 7 |
| Especie del huésped | Rabbit |
| Inmunógeno | regulator of G-protein signaling 7. (Uniprot# P49802) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?