missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
RFTN2 Polyclonal antibody specifically detects RFTN2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | RFTN2 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Janelia Fluor 525 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | C2orf11, chromosome 2 open reading frame 11, FLJ30574, MGC117313, raftlin family member 2, raftlin-2, Raft-linking protein 2 |
| Especie del huésped | Rabbit |
| Inmunógeno | raftlin family member 2. (Uniprot# Q52LD8) |
| Método de purificación | Immunogen Affinity-purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?