missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RCC2 Polyclonal antibody specifically detects RCC2 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | RCC2 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500 - 1:2000, ELISA |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | DKFZp762N0610, KIAA1470, protein RCC2, RCC1-like protein TD-60, regulator of chromosome condensation 2, TD60, TD-60, Telophase disk protein of 60 kDa |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 323-522 of human RCC2 (NP_001129676.1).,, Sequence:, QILPVPNVVVRDVACGANHTLVLDSQKRVFSWGFGGYGRLGHAEQKDEMVPRLVKLFDFPGRGASQIYAGYTCSFAVSEVGGLFFWGATNTSRESTMYPKAVQDLCGWRIRSLACGKSSIIVAADESTISWGPSPTFGELGYGDHKPKSSTAAQEVKTLDGIFSEQVAMGYSHSLVIARDESETEKEKIKKLPEYNPRTL |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?