missing translation for 'onlineSavingsMsg'
Learn More

RBM33 Antibody, R&D Systems™

Código de producto. 18498400 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
25 μL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
18498400 25 μL 25 microlitros
18229396 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18498400 Proveedor Novus Biologicals N.º de proveedor NBP18420025ul
 Artículo restringido
Fecha estimada de envoi: 07-10-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

RBM33 Polyclonal specifically detects RBM33 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno RBM33
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gen DKFZp434D1319, MGC20460, proline-rich protein 8, RNA binding motif protein 33, RNA-binding motif protein 33, RNA-binding protein 33
Símbolos de los genes RBM33
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids:QHPPQHQHHHHHHHLSVPPPPLMPMSQPQFRPHVQTAQPQASSSRMQCPQRQGLRHNTTSQNVSKRPMQQMQPTAPRNSNLREL
Método de purificación Affinity Purified
Cantidad 25 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 155435
Especificidad de la prueba Specificity of human RBM33 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Tipo de producto Antibody
Isotype IgG
Línea de productos Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.