missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RAB3IL1 Monoclonal specifically detects RAB3IL1 in Human/Mouse/Rat samples. It is validated for Flow Cytometry/Western Blot.
Specifications
Specifications
| Antígeno | RAB3IL1 |
| Aplicaciones | Flow Cytometry, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI1B5 |
| Conjugado | mFluor Violet 610 SE |
| Dilución | Flow Cytometry |
| Formulación | 50mM Sodium Borate |
| Alias de gen | GRAB, guanine nucleotide exchange factor for Rab3A, guanine nucleotide exchange factor for Rab-3A, RAB3A interacting protein (rabin3)-like 1, Rab-3A-interacting-like protein 1, Rab3A-interacting-like protein 1, Rabin3-like 1 |
| Especie del huésped | Mouse |
| Inmunógeno | Full length human recombinant protein of human RAB3IL1(NP_037533) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?