missing translation for 'onlineSavingsMsg'
Learn More

PTCD3 Antibody - BSA Free, Novus Biologicals™

Código de producto. 18663481 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mL
0.1 mL
Tamaño de la unidad:
0.01 ml
0.02 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18663481 0.02 mL 0.02 ml
18686180 0.1 mL 0.01 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18663481 Supplier Novus Biologicals Supplier No. NBP2946970.02ml

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTCD3 Polyclonal antibody specifically detects PTCD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno PTCD3
Aplicaciones Western Blot, Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Formulación PBS (pH 7.3), 50% glycerol
Alias de gen DKFZp666K071, FLJ20758, Pentatricopeptide repeat domain 3, pentatricopeptide repeat-containing protein 3, mitochondrial, Transformation-related gene 15 protein, TRG-15
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 450-689 of human PTCD3 (NP_060422.4). PDQHRNFYYSKFFDLICLMEQIDVTLKWYEDLIPSAYFPHSQTMIHLLQALDVANRLEVIPKIWKDSKEYGHTFRSDLREEILMLMARDKHPPELQVAFADCAADIKSAYESQPIRQTAQDWPATSLNCIAILFLRAGRTQEAWKMLGLFRKHNKIPRSELLNELMDSAKVSNSPSQAIEVVELASAFSLPICEGLTQRVMSDFAINQEQKEALSNLTALTSDSDTDSSSDSDSDTSEGK
Método de purificación Affinity purified
Cantidad 0.02 mL
Estado normativo RUO
Disciplina de investigación Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 55037
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.