missing translation for 'onlineSavingsMsg'
Learn More

Protocadherin beta 11 Antibody, Novus Biologicals™

Código de producto. p-200037578 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.1 mL
25ul
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18421092 25ul 25 microlitros
18100340 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18421092 Proveedor Novus Biologicals N.º de proveedor NBP21373825ul

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Protocadherin beta 11 Polyclonal specifically detects Protocadherin beta 11 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Protocadherin beta 11
Aplicaciones Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gen cadherin ME2, ME2, MGC138337, MGC142171, PCDH-beta-11, PCDH-BETA11, protocadherin beta 11, protocadherin beta-11
Símbolos de los genes PCDHB11
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids: KQMLLEIPENSPVGAVFLLESAK
Método de purificación Affinity Purified
Cantidad 25ul
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 56125
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.