Learn More
Novus Biologicals™ Protein kinase-like protein SgK493 Overexpression Lysate
Tienda Bio Techne ProductosDescripción
The Protein kinase-like protein SgK493 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
| Línea de células | The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_612379. The protein contains a C-terminal DDK tag. |
| Para utilizar con (aplicación) | Western Blot |
| Cantidad | 0,1 mg |
| Tampón de almacenamiento | RIPA buffer |
| Pureza | Protein |
| Especificidad de la prueba | Homo sapiens Protein kinase-like protein SgK493, mRNA. |
| Contenido y almacenamiento | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
Para uso exclusivo en investigación