missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Proprotein Convertase 9/PCSK9 Antibody (lodelcizumab) - Humanized, Novus Biologicals™

Product Code. 30048517 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Unit Size:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30048517 50 μg 50 microgramos
30046534 1 mg 1 miligramo
30045511 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30048517 Supplier Novus Biologicals™ Supplier No. NBP32836750ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Proprotein Convertase 9/PCSK9 Monoclonal antibody specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Proprotein Convertase 9/PCSK9
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon lodelcizumab
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen Q8NBP7
Alias de gen EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9
Especie del huésped Human
Inmunógeno PCSK9
Método de purificación Protein A purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cholesterol Metabolism, Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 255738
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.