Learn More
Clorhidrato de propiverina, TRC
Product Code. 30422318
Change view
Click to view available options
Cantidad:
100 mg
500 mg
1 g
Unit Size:
500 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30422318 500 mg 500 miligramos
30414429 1 g -
30417751 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422318 Supplier Toronto Research Chemicals Supplier No. TRCP828000500MG
This item is not returnable. View return policy
Moléculas orgánicas de alta pureza y estándares analíticos, implementados estratégicamente en todo el mundo para impulsar la innovación y el éxito comercial.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54556-98-8
Fórmula molecular C23 H29 N O3 . Cl H
Peso molecular (g/mol) 403.94
Sinónimo α-phenyl-α-propoxy-Benzeneacetic acid 1-methyl-4-piperidinyl ester hydrochloride (1:1), Acetic acid, diphenylpropoxy-, 1-methyl-4-piperidyl ester, hydrochloride (7CI), Benzeneacetic acid, α-phenyl-α-propoxy-, 1-methyl-4-piperidinyl ester, hydrochloride (9CI), Mictonetten, Mictonorm, P 4, P 4 (anticholinergic), Pollarine, Propiverine hydrochloride
Nombre IUPAC (1-metilpiperidina-4-il) 2,2-difenil-2-propoxyaceato; Cloruro
SMILES Cl.CCCOC(C(=O)OC1CCN(C)CC1)(c2ccccc2)c3ccccc3
Specifications
Nombre del producto químico o material Propiverine, Hydrochloride
Notas de grado de pureza HPLC
Cantidad 500 mg
Fórmula InChI InChI=1S/C23H29NO3.ClH/c1-3-18-26-23(19-10-6-4-7-11-19,20-12-8-5-9-13-20)22(25)27-21-14-16-24(2)17-15-21;/h4-13,21H,3,14-18H2,1-2H3;1H
Formula Weight (peso de la fórmula) 403.1914
Porcentaje de pureza >95%