missing translation for 'onlineSavingsMsg'
Learn More

PRAT4A/TNRC5 Antibody, Novus Biologicals™

Código de producto. 18140742 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18140742 0.1 mL 0.10 ml
18148191 0.025 mL 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18140742 Proveedor Novus Biologicals N.º de proveedor NBP223847

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody has been used in 1 publication

PRAT4A/TNRC5 Polyclonal specifically detects PRAT4A/TNRC5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PRAT4A/TNRC5
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin 1: 1000-1:2000
Formulación 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide
N.º de referencia del gen Q9BT09
Alias de gen CAG4A, canopy 3 homolog (zebrafish), CTG4A, ERDA5CTG repeat protein 4a, Expanded repeat-domain protein CAG/CTG 5, PRAT4ACAG repeat containing, Protein associated with TLR4, protein associated with Toll-like receptor 4A, protein canopy homolog 3, TNRC5expanded repeat domain, CAG/CTG 5, trinucleotide repeat containing 5, Trinucleotide repeat-containing gene 5 protein
Símbolos de los genes CNPY3
Especie del huésped Rabbit
Inmunógeno A portion of amino acids 200-250 of human PRAT4A was used as the immunogen for this antibody.
Método de purificación Unpurified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10695
Especificidad de la prueba The amino acid sequence used as immunogen is 100% homologous in mouse and rat.
Especies diana Human
Contenido y almacenamiento Store at -20C. Avoid freeze-thaw cycles.
Formulario Antisera
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.