missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
PNO1 Polyclonal antibody specifically detects PNO1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Especificaciones
Especificaciones
| Antígeno | PNO1 |
| Aplicaciones | ELISA, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | mFluor Violet 610 SE |
| Formulación | 50mM Sodium Borate |
| Alias de gen | KHRBP1, KH-type RNA binding protein 1, KH-type RNA-binding protein 1, partner of NOB1 homolog (S. cerevisiae), RNA binding protein, RNA-binding protein PNO1, RRP20 |
| Especie del huésped | Rabbit |
| Inmunógeno | partner of NOB1 homolog. (Uniprot# Q9NRX1) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?