missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
PIR-A/B Monoclonal antibody specifically detects PIR-A/B in Mouse samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
Especificaciones
| Antígeno | PIR-A/B |
| Aplicaciones | Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 404127 |
| Conjugado | mFluor Violet 610 SE |
| Dilución | Flow Cytometry, CyTOF-ready |
| Formulación | 50mM Sodium Borate |
| Especie del huésped | Rat |
| Inmunógeno | Mouse myeloma cell line NS0-derived recombinant mouse PIR-A1, Gly24-Met650, Accession # A2NTJ8 |
| Método de purificación | Protein A or G purified from hybridoma culture supernatant |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?