missing translation for 'onlineSavingsMsg'
Learn More

PIEZO1 Antibody (1081418) [PerCP], Novus Biologicals™

Código de producto. 30679149 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30679149 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30679149 Proveedor Novus Biologicals N.º de proveedor FAB11557C

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

PIEZO1 Monoclonal antibody specifically detects PIEZO1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno PIEZO1
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Clon 1081418
Conjugado PerCP
Formulación PBS
Alias de gen FAM38A, member A, membrane protein induced by beta-amyloid treatment, Mib, piezo-type mechanosensitive ion channel component 1
Especie del huésped Mouse
Inmunógeno PIEZO1 containing synthetic peptide, Accession # Q92508
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Alzheimers Research, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 9780
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.