missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
PI4KB/PI4KIII beta Polyclonal specifically detects PI4KB/PI4KIII beta in Human samples. It is validated for Immunoprecipitation, Western Blot.
Especificaciones
Especificaciones
| Antígeno | PI4KB/PI4KIII beta |
| Aplicaciones | Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 647 |
| Dilución | Western Blot, Immunoprecipitation |
| Formulación | 50mM Sodium Borate |
| Alias de gen | DKFZp686O1820, EC 2.7.1, EC 2.7.1.67, FLJ30129, phosphatidylinositol 4-kinase, catalytic, beta, pi4K92, PI4Kbeta, PI4K-beta, PI4KIIIBETA, type III phosphatidylinositol 4-kinase beta, wortmannin-sensitive |
| Especie del huésped | Rabbit |
| Inmunógeno | The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Phosphatidylinositol 4-Kinase, Catalytic, beta using the numbering given in entry CAA09495.1 (GeneID 5298). |
| Método de purificación | Immunogen affinity purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?