missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PHF21A Polyclonal specifically detects PHF21A in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antígeno | PHF21A |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 750 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | BHC80a, BHC80BM-006, BRAF35-HDAC complex protein BHC80, KIAA1696BRAF35/HDAC2 complex (80 kDa), PHD finger protein 21A |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PHF21A. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?