missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
PEX5L Polyclonal antibody specifically detects PEX5L in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Especificaciones
Especificaciones
| Antígeno | PEX5L |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | DyLight 680 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | PEX5-related protein, peroxisomal biogenesis factor 5-like, PEX5R, PEX5RP, PXR2, PXR2B |
| Especie del huésped | Rabbit |
| Inmunógeno | peroxisomal biogenesis factor 5-like. (Uniprot# Q8IYB4) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?