Descripción
Pea3 Polyclonal antibody specifically detects Pea3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Especificaciones
Especificaciones
| Antígeno | Pea3 |
| Aplicaciones | Western Blot, ELISA |
| Clasificación | Polyclonal |
| Conjugado | DyLight 680 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | Adenovirus E1A enhancer-binding protein, E1A-FE1A enhancer binding protein, E1AFETS translocation variant 4, ets variant 4, ets variant gene 4 (E1A enhancer binding protein, E1AF), ets variant gene 4 (E1A enhancer-binding protein, E1AF), EWS protein/E1A enhancer binding protein chimera, PEA3, PEAS3, Polyomavirus enhancer activator 3 homolog, polyomavirus enhancer activator-3, Protein PEA3 |
| Especie del huésped | Rabbit |
| Inmunógeno | Pea3 antibody was raised against a peptide corresponding to 19 amino acids near the amino terminus of human Pea3. Amino Acid Squence: HSPTTRIKKEPQSPRTDPA |
| Método de purificación | Peptide affinity purified |
| Cantidad | 0.1 mL |
| Mostrar más |