missing translation for 'onlineSavingsMsg'
Learn More

PDCD10 Antibody (2A8) [CoraFluorÖ 1], Novus Biologicals Biologicals™

Código de producto. 30519828 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad
30519828 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Product Code. 30519828 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP343064CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDCD10 Monoclonal antibody specifically detects PDCD10 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno PDCD10
Aplicaciones ELISA, Immunohistochemistry, Western Blot
Clasificación Monoclonal
Clon 2A8
Conjugado CoraFluor 1
Formulación PBS
Alias de gen apoptosis-related protein 15, cerebral cavernous malformations 3, MGC1212, MGC24477, programmed cell death 10, programmed cell death protein 10, TF-1 cell apoptosis-related protein 15, TFAR15CCM3Cerebral cavernous malformations 3 protein
Especie del huésped Mouse
Inmunógeno programmed cell death 10 - (Uniprot# Q9BUL8)
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology, Neuroscience, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 11235
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Tipo de producto Antibody
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.