Learn More
p16INK4a/CDKN2A Antibody, Novus Biologicals™
Código de producto. 18624136 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
18624136 100 μg 100 microlitros
18628125 50 μg 50 microlitros
2 options
Código de producto. 18624136
Marca: Novus Biologicals NBP298881100ul
Rabbit Polyclonal Antibody
p16INK4a/CDKN2A Polyclonal antibody specifically detects p16INK4a/CDKN2A in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno p16INK4a/CDKN2A
Aplicaciones Western Blot, Immunofluorescence, Immunoprecipitation
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunoprecipitation 1-4 μL/mg of lysate
Formulación 0.2 um filtered solution in PBS
Alias de gen ARF, CDK4 inhibitor p16-INK4, CDK4IP14ARF, CDKN2, cell cycle negative regulator beta, CMM2P16-INK4A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2A, cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4), INK4, INK4A, MLMP16INK4, MTS-1, MTS1P14, Multiple tumor suppressor 1, p14, p14ARF, p16, p16-INK4, p16INK4a, p16-INK4a, P19, p19ARF, TP16
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human p16INK4a/CDKN2A.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación ABC Transporters, Biologically Active Proteins, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, MAP Kinase Signaling, Neuroscience, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 1029
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG