Descripción
These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.
Especificaciones
Especificaciones
| Reactividad cruzada | Human |
| Tipo | Ovary Tissue Slides |
| Estado normativo | This product is for research use only and is not approved for use in humans or in clinical diagnosis. |
| Especie | Humano |
| Línea de productos | Novus |