missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ODF2L Polyclonal antibody specifically detects ODF2L in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | ODF2L |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | PE |
| Formulación | PBS |
| Alias de gen | dJ977L11.1, KIAA1229RP5-977L11.1, MGC111060, outer dense fiber of sperm tails 2-like, outer dense fiber protein 2-like |
| Especie del huésped | Rabbit |
| Inmunógeno | outer dense fiber of sperm tails 2-like. (Uniprot# Q9ULJ1) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?