Learn More
Invitrogen™ O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (RL2)
GREENER_CHOICE
Código de producto. 11518842
Cambiar vista
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
11518842 100 μL 100 microlitros
1 options
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:27969
Código de producto. 11518842 Proveedor Invitrogen™ N.º de proveedor MA1072
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:27969
Mouse Monoclonal Antibody
MA1-072 detects nuclear pore complex (NPC), cytoplasmic and intranuclear O-linked glycoproteins from human, mouse, virus and rat tissues. MA1-072 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody recognizes up to eight different proteins from the NPC of approximately 210, 180, 145, 100, 63, 58, 54 and 45 kDa as well as other O-linked glycoproteins outside of the NPC. Immunofluorescence staining of the NPC with MA1-072 primarily labels NPC O-linked glycoproteins and has been successfully used on a wide variety of mammalian cells. Labeling occurs at the NPC, with most of the labeling at the cytoplasmic and/or nucleoplasmic margins, as well as within the nucleus. The MA1-072 immunogen is pore complex-lamina fraction purified from rat liver nuclear envelopes.
O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno O-linked N-acetylglucosamine (O-GlcNAc)
Aplicaciones ChIP Assay, Dot Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon RL2
Concentración 2 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.05% sodium azide
génica OGT
N.º de referencia del gen O15294, Q8CGY8, P56558
Alias de gen GlcNAc; HINCUT-1; HRNT1; O-GLCNAC; O-GlcNAc transferase p110 subunit; O-GlcNAc transferase subunit p110; OGT; O-linked N-acetylglucosamine (GlcNAc) transferase; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase 110 kDa subunit; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit; uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyl transferase
Símbolos de los genes OGT
Especie del huésped Mouse
Inmunógeno Pore complex-lamina fraction purified from rat liver nuclear envelopes.
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 108155, 26295, 8473
Especies diana Chemical
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Molécula diana Chemical