Learn More
NVS-ZP7-4, MedChemExpress
Código de producto. 16475616
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16475616 100 mg 100 miligramos
16443256 25 mg 25 miligramos
16431087 50 mg 50 miligramos
16424828 10 mg 10 miligramos
16417577 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16475616 Proveedor Medchem Express N.º de proveedor HY114395
MedChemExpress NVS-ZP7-4 is a Zinc transporter SLC39A7 (ZIP7) inhibitor that is also the first reported chemical tool to probe the impact of modulating ER zinc levels and investigate ZIP7 as a novel druggable node in the Notch pathway.
NVS-ZP7-4 is a Zinc transporter SLC39A7 (ZIP7) inhibitor that is also the first reported chemical tool to probe the impact of modulating ER zinc levels and investigate ZIP7 as a novel druggable node in the Notch pathway.
Especificaciones
Nombre del producto químico o material NVS-ZP7-4
CAS 2349367-89-9
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C28H28FN5OS
Cantidad 100 mg
Información de solubilidad DMSO : ≥ 125 mg/mL (249.19 mM)
SMILES O=C(NC12CCN(C[C@@H](NC3=NC4=CC=C(F)C=C4S3)CC5=CC=CC=C5)CC2)NC6=C1C=CC=C6
Peso molecular (g/mol) 501.62
Formula Weight (peso de la fórmula) 501.62
Porcentaje de pureza 98.68%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation