missing translation for 'onlineSavingsMsg'
Learn More

Nudel Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™

Código de producto. 30505708 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30505708 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30505708 Proveedor Novus Biologicals N.º de proveedor NBP338185JF669

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Nudel Polyclonal antibody specifically detects Nudel in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Nudel
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Janelia Fluor 669
Formulación 50mM Sodium Borate
Alias de gen DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 1-345 of human Nudel (NP_110435.1).,, Sequence:, MDGEDIPDFSSLKEETAYWKELSLKYKQSFQEARDELVEFQEGSRELEAELEAQLVQAEQRNRDLQADNQRLKYEVEALKEKLEHQYAQSYKQVSVLEDDLSQTRAIKEQLHKYVRELEQANDDLERAKRATIVSLEDFEQRLNQAIERNAFLESELDEKESLLVSVQRLKDEARDLRQELAVRERQQEVTRKSAPSSPTLDCEKMDSAVQASLSLPATPVGKGTENTFPSPKAIPNGFGTSPLTPSARISALNIVGDLLRKVGALESKLAACRNFAKDQASRKSYISGNVNCGVLNGNGTKFSRSGHTSFFDKGAVNGFDPAPPPPGLGSSRPSSAPGMLPLSV
Método de purificación Affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 81565
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Tipo de producto Antibody
Formulario Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.