Descripción
NT-proBNP Monoclonal antibody specifically detects NT-proBNP in Human samples. It is validated for Western Blot, ELISA
Especificaciones
Especificaciones
| Antígeno | NT-proBNP |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | 11D1cc |
| Conjugado | DyLight 594 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein |
| Especie del huésped | Mouse |
| Inmunógeno | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic NT-proBNP, corresponding to a.a.r. 28-45, conjugated with carrier protein. |
| Método de purificación | Protein A purified |
| Mostrar más |