missing translation for 'onlineSavingsMsg'
Learn More

Nrip2 Antibody, Novus Biologicals™

Código de producto. p-200044273 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
25ul
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18468940 25ul 25 microlitros
18292848 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18468940 Proveedor Novus Biologicals N.º de proveedor NBP18503325ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody has been used in 1 publication

Nrip2 Polyclonal specifically detects Nrip2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Nrip2
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
N.º de referencia del gen Q9BQI9
Alias de gen DKFZP761G1913, nuclear receptor interacting protein 2, nuclear receptor-interacting protein 2
Símbolos de los genes NRIP2
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids:PTPPAGAMSTKQEARRDEGEARTRGQEAQLRDRAHLSQQRRLKQATQFLHK
Método de purificación Affinity Purified
Cantidad 25ul
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 83714
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.