Learn More
Noxa Antibody, Novus Biologicals™
Código de producto. 18706953 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.025 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18706953 0.025 mg 25 microgramos
18796523 - 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18706953 Proveedor Novus Biologicals N.º de proveedor NB10056368SS
Rabbit Polyclonal Antibody has been used in 5 publications
Noxa Polyclonal specifically detects Noxa in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Noxa
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1 - 2 ug/mL, Immunohistochemistry 1:50 - 1:250, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:50 - 1:250. Use reported in scientific literature (Inta et al. (2006))
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen Q9JM54
Alias de gen adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
Símbolos de los genes PMAIP1
Especie del huésped Rabbit
Inmunógeno This Noxa antibody was developed by immunizing rabbits with a mixture of synthetic peptides containing amino acids 51-66 and 75-90 of mouse Noxa.
Peso molecular del antígeno 11.6 kDa
Método de purificación Protein G purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 5366
Especificidad de la prueba Amino acids 51-66 of mouse Noxa are 100% homologous in mouse and 77% in rat; amino acids 75-90 of mouse Noxa are 100% homologous in mouse, 93% homologous in rat and 76% homologous in human.
Especies diana Human, Mouse, Mouse
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only