missing translation for 'onlineSavingsMsg'
Learn More

NME3 Antibody [Alexa Fluor™ 405], Novus Biologicals™

Código de producto. 30606386 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30606386 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30606386 Proveedor Novus Biologicals N.º de proveedor NBP347331AF405

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NME3 Polyclonal antibody specifically detects NME3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno NME3
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Alexa Fluor 405
Formulación 50mM Sodium Borate
Alias de gen c371H6.2, DR-nm23NDPK-C, EC 2.7.4.6, KIAA0516, NDK 3, NDP kinase 3, NDP kinase C, NDPKC, NM23H3, NM23-H3, non-metastatic cells 3, protein expressed in, nucleoside diphosphate kinase 3, Nucleoside diphosphate kinase C
Especie del huésped Rabbit
Inmunógeno non-metastatic cells 3, protein expressed in. (Uniprot# Q13232)
Método de purificación Immunogen Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 4832
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.