Learn More
NEDD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13993-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody works well in WB, IHC, IP and IF application.
Neural precursor cell expressed developmentally down-regulated protein 1 (NEDD1) is required for mitosis progression. NEDD1 promotes the nucleation of microtubules from the spindle.Specifications
NEDD1 | |
Polyclonal | |
Unconjugated | |
NEDD1 | |
GCP WD, NEDD 1, NEDD1, Protein NEDD1, TUBGCP7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
121441, 17997, 299730 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P33215, Q8NHV4 | |
NEDD1 | |
NEDD1 Fusion Protein Ag5077 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NEDD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction