missing translation for 'onlineSavingsMsg'
Learn More

NDUFA3 Antibody, Novus Biologicals™

Código de producto. 18622502 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Unit Size:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18622502 50 μg 50 microlitros
18661702 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18622502 Supplier Novus Biologicals Supplier No. NBP29815250ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDUFA3 Polyclonal antibody specifically detects NDUFA3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno NDUFA3
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin 1:50-1:200
Formulación PBS (pH 7.0)
Alias de gen B9, CI-B9, complex I B9 subunit, Complex I-B9, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3 (9kD, B9), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3, 9kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3, NADH-ubiquinone oxidoreductase B9 subunit
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human NDUFA3 fragment.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 4696
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.