missing translation for 'onlineSavingsMsg'
Learn More

NDUFA12 Antibody - BSA Free, Novus Biologicals™

Código de producto. 18653592 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mL
0.1 mL
Tamaño de la unidad:
0.02 ml
0.10 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18653592 0.02 mL 0.02 ml
18692372 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18653592 Proveedor Novus Biologicals N.º de proveedor NBP2940420.02ml

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

NDUFA12 Polyclonal antibody specifically detects NDUFA12 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno NDUFA12
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500 - 1:2000
Formulación PBS (pH 7.3), 50% glycerol
Alias de gen 13 kDa differentiation-associated protein, B17.2,13kDa differentiation-associated protein, CIB17.2, CI-B17.2, complex I B17.2 subunit, Complex I-B17.2, DAP13NADH-ubiquinone oxidoreductase subunit B17.2, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12, NADH: ubiquinone oxidoreductase
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human NDUFA12 (NP_061326.1). MELVQVLKRGLQQITGHGGLRGYLRVFFRTNDAKVGTLVGEDKYGNKYYEDNKQFFGRHRWVVYTTEMNGKNTFWDVDGSMVPPEWHRWLHSMTDDPPTTKPLTARKFIWTNHKFNVTGTPEQYVPYSTTRKKIQEWIPPSTPYK
Método de purificación Affinity purified
Cantidad 0.02 mL
Estado normativo RUO
Disciplina de investigación Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 55967
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.