missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse M-CSF, Animal-Free Recombinant Protein, PeproTech®

Código de producto. 17886608
Change view
Click to view available options
Cantidad:
250 μg
2 μg
50 μg
Tamaño de la unidad:
2 microgramos
250 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17886608 50 μg 50 microgramos
17806818 250 μg 250 microgramos
17826578 2 μg 2 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17886608 Proveedor Gibco™ N.º de proveedor AF3150250UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Murine M-CSF is a 36.4 kDa homodimeric protein consisting of two 156 amino acid polypeptide subunits. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P07141
Para utilizar con (aplicación) Functional Assay
Formato Lyophilized
Formulación protein with no preservative
ID de gen (Entrez) 12977
Peso molecular 36.4 kDa
Nombre Mouse M-CSF, Animal-Free
Método de purificación HPLC, SDS-PAGE
Cantidad 50 μg
Estado normativo RUO
Concentración de endotoxinas <0.1 EU/ μg
Alias de gen C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1
Nombre común M-CSF
Símbolo de gen Csf1
Actividad biológica Determined by its ability to stimulate the proliferation of murine M-NFS-60 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
Contenido y almacenamiento -20°C
Actividad Determined by its ability to stimulate the proliferation of murine M-NFS-60 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.