missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MLK4, GST Tag Recombinant Protein

Código de producto. 17168795
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17168795 1 mg 1 miligramo
15970303 10 μg 10 microgramos
17198745 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17168795 Proveedor Invitrogen™ N.º de proveedor A33382

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This MLK4 recombinant human protein (114-420) was expressed in insect cells. MLK4 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for MLK4 is NP_115811.2.

MLK4 or mixed lineage kinase 4 belongs to the superfamily of MAP kinase kinases (MAP3K1) which contain both ser/thr and tyr kinases activity in their catalytic domains. The structure of this kinase family incorporates an N-terminal Src homology (SH3) domain, followed by the kinase domain, a leucine zipper region, and a CDC42 /RAC -interactive binding (CRIB) motif, but divergent C-terminal regions. MLK4 is highly expressed in kidney and pancreas. MLK4 is a negative regulator of TLR4 and does not activate JNK1/MAPK8 pathway, p38/MAPK14, nor ERK2/MAPK1 pathways.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso Q5TCX8
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris HCl with 0.1mM PMSF, 150mM NaCl, 10mM glutathione, 25% glycerol, 0.25mM DTT, 0.1mM EDTA and no preservative; pH 7.5
ID de gen (Entrez) 84451
Peso molecular 60.5 kDa
Nombre Human MLK4, GST Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 1 mg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen BC021891; cDNA sequence BC021891; dJ862P8.3; Kiaa1804; MAP3K21; Mitogen-activated protein kinase kinase kinase 21; Mitogen-activated protein kinase kinase kinase MLK4; mixed lineage kinase 4; mKIAA1804; MLK4; RGD1306091; similar to Mixed lineage kinase 4
Nombre común MLK4
Símbolo de gen MAP3K21
Familia Quinasa MAP Kinases
Longitud de la proteína 114-420
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia 114-420
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Condiciones de envío Dry Ice
Sistema de expresión Insect cells
Forma de proteína Recombinant, Active, Catalytic
Formulario Liquid
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.