Learn More
ML216, MedChemExpress
Código de producto. 16497827
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16497827 5 mg 5 miligramos
16465108 10 mg 10 miligramos
16433296 25 mg 25 miligramos
16431347 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16497827 Proveedor Medchem Express N.º de proveedor HY12342
MedChemExpress ML216 (CID-49852229) is a potent, selective and cell permeable inhibitor of the DNA unwinding activity of BLM helicase with IC50s of 2.98 μM and 0.97 μM for BLMfull-length and BLM636-1298, respectively. ML216 inhibits ssDNA-dependent ATPase activity of BLM with a Ki of 1.76 μM. Antitumor avtivity.
ML216 (CID-49852229) is a potent, selective and cell permeable inhibitor of the DNA unwinding activity of BLM helicase with IC50s of 2.98 μM and 0.97 μM for BLMfull-length and BLM636-1298, respectively. ML216 inhibits ssDNA-dependent ATPase activity of BLM with a Ki of 1.76 μM. Antitumor avtivity.
Especificaciones
Nombre del producto químico o material ML216
CAS 1430213-30-1
Color Deep Yellow
Notas de grado de pureza Research
Fórmula molecular C15H9F4N5OS
Cantidad 5 mg
Sinónimo CID-49852229
Información de solubilidad DMSO : 20 mg/mL (52.18 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(F)C(C(F)(F)F)=C1)NC2=NN=C(S2)C3=CC=NC=C3
Peso molecular (g/mol) 383.32
Formula Weight (peso de la fórmula) 383.32
Porcentaje de pureza 98.07%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death