missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MIPOL1 Polyclonal antibody specifically detects MIPOL1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antígeno | MIPOL1 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Biotin |
| Formulación | PBS |
| Alias de gen | DKFZp313M2036, MGC34010, mirror-image polydactyly 1, mirror-image polydactyly gene 1 protein |
| Especie del huésped | Rabbit |
| Inmunógeno | mirror-image polydactyly 1. (Uniprot# Q8TD10) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?