Learn More
MHC class II (I-A/I-E) Antibody (M5/114.15.2) - BSA Free, Novus Biologicals™
Código de producto. 18008405 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.05 mg
0.1 mg
0.5 mg
Tamaño de la unidad:
0.05 miligramo
0.10 miligramo
0.50 miligramo
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
18008405 0.05 mg 0.05 miligramo
18038405 0.1 mg 0.10 miligramo
18751174 0.5 mg 0.50 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 18008405 Proveedor Novus Biologicals N.º de proveedor NBP1433120.05MG
Rat Monoclonal Antibody has been used in 11 publications
MHC class II (I-A/I-E) Monoclonal specifically detects MHC class II (I-A/I-E) in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno MHC Class II
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Clasificación Monoclonal
Clon M5/114.15.2
Concentración 0.5 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:100-1:2000, Flow Cytometry < 0.125 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulación Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Alias de gen class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Símbolos de los genes HLA-DMA
Especie del huésped Rat
Inmunógeno The immunogen for this antibody was MHC Class II.
Método de purificación Affinity Purified
Cantidad 0.05 mg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primario o secundario Primary
ID de gen (Entrez) 3108
Especificidad de la prueba The monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes.
Especies diana Mouse
Contenido y almacenamiento Store at 4C. Do not freeze.
Isotype IgG2b κ
Mostrar más Mostrar menos
For Research Use Only