missing translation for 'onlineSavingsMsg'
Learn More

MafB Antibody (372239) [DyLight 755], Novus Biologicals™

Código de producto. 30632377 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30632377 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30632377 Proveedor Novus Biologicals N.º de proveedor FAB3810Z

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MafB Monoclonal antibody specifically detects MafB in Human,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno MafB
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon 372239
Conjugado DyLight 755
Formulación 50mM Sodium Borate
Alias de gen Kreisler (mouse) maf-related leucine zipper homolog, Kreisler maf-related leucine zipper homolog, KRMLMAFB/Kreisler basic region/leucine zipper transcription factor, Maf-B, MGC43127, transcription factor MafB, V-maf musculoaponeurotic fibrosarcoma oncogene homolog B, v-maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian)
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human MafB, Met1-Leu323, Accession # Q9Y5Q3
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 9935
Especies diana Human, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.