Learn More
Linrodostat, MedChemExpress
Código de producto. 16476137
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Tamaño de la unidad:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16476137 5 mg 5 miligramos
16469936 50 mg 50 miligramos
16413418 10 mg 10 miligramos
16412766 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476137 Supplier Medchem Express Supplier No. HY101560
This item is not returnable. View return policy
MedChemExpress Linrodostat (BMS-986205) is a selective and irreversible indoleamine 2,3-dioxygenase 1 (IDO1) inhibitor with an IC50 value of 1.1 nM in IDO1-HEK293 cells. Linrodostat is well tolerated with potent pharmacodynamic activity in advanced cancers.
Linrodostat (BMS-986205) is a selective and irreversible indoleamine 2,3-dioxygenase 1 (IDO1) inhibitor with an IC50 value of 1.1 nM in IDO1-HEK293 cells. Linrodostat is well tolerated with potent pharmacodynamic activity in advanced cancers.
Specifications
Nombre del producto químico o material Linrodostat
CAS 1923833-60-6
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C24H24ClFN2O
Cantidad 5 mg
Sinónimo BMS-986205 ONO-7701
Información de solubilidad DMSO : 50 mg/mL (121.68 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(Cl)C=C1)[C@H](C)[C@@](CC2)([H])CC[C@H]2C3=CC=NC4=C3C=C(F)C=C4
Peso molecular (g/mol) 410.91
Formula Weight (peso de la fórmula) 410.91
Porcentaje de pureza 98.04%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death