missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX2 Antibody (CL6137) [Alexa Fluor« 488], Novus Biologicals Biologicals™
Tienda Bio Techne ProductosDescripción
LHX2 Monoclonal antibody specifically detects LHX2 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | LHX2 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CL6137 |
| Conjugado | Alexa Fluor 488 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | hLhx2, Homeobox protein LH-2, LH-2, LH2MGC138390, LIM homeobox 2, LIM homeobox protein 2, LIM HOX gene 2, LIM/homeobox protein Lhx2 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was generated using a synthetic peptide of P50458, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?