missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LDLRAD1 Polyclonal antibody specifically detects LDLRAD1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antígeno | LDLRAD1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Janelia Fluor 525 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | low density lipoprotein receptor A domain containing 1, low density lipoprotein receptor class A domain containing 1, low-density lipoprotein receptor class A domain-containing protein 1 |
| Especie del huésped | Rabbit |
| Inmunógeno | low density lipoprotein receptor class A domain containing 1. (Uniprot# Q5T700) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?