missing translation for 'onlineSavingsMsg'
Learn More

LAMP-1/CD107a Antibody, DyLight 594, Novus Biologicals™

Código de producto. 18498484 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
18498484 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18498484 Proveedor Novus Biologicals N.º de proveedor NB600956DL594
  • ONLINE EXCLUSIVE PRICE
    617.00€
    582.75€ / 0.1 ml
    Ahorro 34.25€
    5% Descuento
 Artículo restringido
Fecha estimada de envoi: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LAMP-1/CD107a Polyclonal specifically detects LAMP-1/CD107a in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno LAMP-1/CD107a
Aplicaciones Immunocytochemistry/Immunofluorescence, Western Blot
Clasificación Polyclonal
Conjugado DyLight 594
Formulación 50mM Sodium Borate
Alias de gen CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1
Especie del huésped Rabbit
Inmunógeno This antibody was generated with a synthetic peptide corresponding to amino acid residues 405-416 of human LAMP1 with N-terminal added cysteine-glycine, conjugated to KLH.
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites
Primario o secundario Primary
ID de gen (Entrez) 3916
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.