Learn More
Lamin B1 Antibody (CL3929), Novus Biologicals™
Código de producto. 18087103 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
25 μL
Tamaño de la unidad:
100 microlitros
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18087103 100 μL 100 microlitros
18601708 25 μL 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18087103 Proveedor Novus Biologicals N.º de proveedor NBP259783
Mouse Monoclonal Antibody has been used in 5 publications
Lamin B1 Monoclonal specifically detects Lamin B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Lamin B1
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 4001
Conjugado Unconjugated
Dilución Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulación PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Alias de gen ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419
Símbolos de los genes LMNB1
Especie del huésped Mouse
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: RTTRGKRKRVDVEESEASSSVSISHSASA
Método de purificación Protein A purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Stem Cell Markers, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 4001
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1
Mostrar más Mostrar menos
For Research Use Only (RUO)