missing translation for 'onlineSavingsMsg'
Learn More

Kv3.4 Antibody [Alexa Fluor™ 647], Novus Biologicals™

Código de producto. 30603672 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30603672 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30603672 Proveedor Novus Biologicals N.º de proveedor NBP346434AF647

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Kv3.4 Polyclonal antibody specifically detects Kv3.4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Kv3.4
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Alexa Fluor 647
Formulación 50mM Sodium Borate
Alias de gen HKSHIIIC, K+ channel subunit, KSHIIIC, Kv3.4, MGC126818, potassium voltage-gated channel subfamily C member 4, potassium voltage-gated channel, Shaw-related subfamily, member 4, Voltage-gated potassium channel subunit Kv3.4
Especie del huésped Rabbit
Inmunógeno potassium voltage-gated channel, Shaw-related subfamily, member 4. (Uniprot# Q03721)
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Alzheimers Research, Neurodegeneration, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 3749
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.