missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
KLHL28 Polyclonal antibody specifically detects KLHL28 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antígeno | KLHL28 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 700 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | BTB (POZ) domain containing 5, BTB/POZ domain-containing protein 5, FLJ20081, kelch-like 28 (Drosophila) |
| Especie del huésped | Rabbit |
| Inmunógeno | kelch-like 28(Drosophila). (Uniprot# Q9NXS3) |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?