Learn More
JNJ-37822681 dihydrochloride, MedChemExpress
Código de producto. 16497117
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
Tamaño de la unidad:
5 miligramos
16497117 5 mg 5 miligramos
1 options
Código de producto. 16497117 Proveedor Medchem Express N.º de proveedor HY111066A
MedChemExpress JNJ-37822681 dihydrochloride is a potent, specific, centrally active, fast-dissociating dopamine D2 receptor antagonist with a moderate binding affinity for the dopamine D2L receptor (Ki =158 nM), which has potential for the treatment of schizophrenia and bipolar disorder.
JNJ-37822681 dihydrochloride is a potent, specific, centrally active, fast-dissociating dopamine D2 receptor antagonist with a moderate binding affinity for the dopamine D2L receptor (Ki =158 nM), which has potential for the treatment of schizophrenia and bipolar disorder.
Especificaciones
Nombre del producto químico o material JNJ-37822681 dihydrochloride
CAS 2108806-02-4
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C17H19Cl2F5N4
Cantidad 5 mg
Información de solubilidad DMSO : 33.33 mg/mL (74.86 mM; Need ultrasonic)
SMILES FC(C1=NN=C(NC2CCN(CC3=CC=C(F)C(F)=C3)CC2)C=C1)(F)F.Cl.Cl
Peso molecular (g/mol) 445.26
Formula Weight (peso de la fórmula) 445.26
Porcentaje de pureza 98.0%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Neuroscience-Neuromodulation