missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 3/CD61 Antibody (Y2/51), Alexa Fluor™ 488, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Especificaciones
Especificaciones
| Antígeno | Integrin beta 3/CD61 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Clasificación | Monoclonal |
| Clon | Y2/51 |
| Conjugado | Alexa Fluor 488 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa |
| Especie del huésped | Mouse |
| Inmunógeno | Phytohemagglutinin (PHA)-stimulated peripheral blood cells |
| Método de purificación | Protein A or G purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?