missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 4/CD49d Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18498854 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad Unit Size
18498854 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18498854 Supplier Novus Biologicals Supplier No. NBP177333AF405
  • ONLINE EXCLUSIVE PRICE
    630.00€
    595.35€ / 0.1 ml
    Ahorro 34.65€
    5% Descuento

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Integrin alpha 4/CD49d Polyclonal antibody specifically detects Integrin alpha 4/CD49d in Human, Mouse, Rat, Insect samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Integrin alpha 4/CD49d
Aplicaciones Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Alexa Fluor 405
Dilución Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulación 50mM Sodium Borate
Alias de gen 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Integrin alpha 4. The immunogen is located within amino acids 220 - 270 of Integrin alpha 4. Amino Acid Squence: TTNKYKAFLDKQNQV
Método de purificación Peptide affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Microautophagy, Neuronal Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 3676
Especies diana Human, Mouse, Rat, Insect, Primate
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.