missing translation for 'onlineSavingsMsg'
Learn More

IMUP Antibody, Novus Biologicals™

Código de producto. 18160152 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
25ul
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18160152 0.1 mL 0.10 ml
18401081 25ul 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18160152 Proveedor Novus Biologicals N.º de proveedor NBP214392

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

IMUP Polyclonal specifically detects IMUP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IMUP
Aplicaciones Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gen C19orf33, chromosome 19 open reading frame 33, H2RSP, HAI-2 related small protein, HAI-2-related small protein, Hepatocyte growth factor activator inhibitor type 2-related small protein, immortalization up-regulated protein, immortalization-upregulated protein, IMUP, IMUP-1, IMUP-2, MGC39135, MGC75180
Símbolos de los genes C19ORF33
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids: SSSDSDTDVKSHAAGSKQHESIPGKAKKPKVKKKEKGKK
Método de purificación Affinity Purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 64073
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.